The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001064.3 | NP_001055.1 | 7 - 8 | 1120 | 80 |
NM_001135055.2 | NP_001128527.1 | 7 - 8 | 1120 | 80 | |
NM_001258028.1 | NP_001244957.1 | 8 - 9 | 1144 | 80 | |
NR_047580.1 | 7 - 8 | 1010 | 80 | ||
XM_011534054.1 | XP_011532356.1 | 8 - 9 | 1082 | 80 | |
XM_011534055.1 | XP_011532357.1 | 8 - 9 | 1169 | 80 | |
GenBank mRNA | AK057092.1 | - | 1 - 2 | 949 | 80 |
AK093667.1 | - | 6 - 7 | 779 | 80 | |
AK223614.1 | - | 7 - 8 | 1028 | 80 | |
AK289454.1 | - | 7 - 8 | 996 | 80 | |
AK293438.1 | - | 8 - 9 | 1052 | 80 | |
AK301231.1 | - | 7 - 8 | 918 | 80 | |
AK303191.1 | - | 8 - 9 | 987 | 80 | |
BC002433.2 | - | 8 - 9 | 909 | 80 | |
BC008615.1 | - | 2 - 3 | 264 | 80 | |
BC009970.2 | - | 7 - 8 | 1018 | 80 | |
BC024026.2 | - | 5 - 6 | 689 | 80 | |
BX647248.1 | - | 7 - 8 | 1006 | 80 | |
BX649193.1 | - | 1 - 2 | 4257 | 80 | |
EU794667.1 | - | 7 - 8 | 1018 | 80 | |
GQ472197.1 | - | 2 - 3 | 264 | 80 | |
KU178361.1 | - | 7 - 8 | 948 | 80 | |
L12711.1 | - | 7 - 8 | 1046 | 80 | |
U55017.1 | - | 7 - 8 | 948 | 80 | |
X67688.1 | - | 8 - 9 | 1018 | 80 |