Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STXBP1
Entrez Gene ID
6812
Gene Name
syntaxin binding protein 1
Gene Aliases
MUNC18-1, NSEC1, P67, RBSEC1, UNC18
Chromosome Location
Chr.9: 127612207 - 127692716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK094794.1 - 1 - 1 599 60

Gene Symbol
MIR3911
Entrez Gene ID
100500872
Gene Name
microRNA 3911
Gene Aliases
mir-3911
Chromosome Location
Chr.9: 127690687 - 127690795 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_037473.1 1 - 1 60

View Full Product Details