The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005164.3 | NP_005155.1 | 8 - 9 | 2306 | 68 |
XM_011538027.2 | XP_011536329.1 | 9 - 10 | 2246 | 68 | |
XM_017018992.1 | XP_016874481.1 | 8 - 9 | 2306 | 68 | |
GenBank mRNA | AJ000327.1 | - | 8 - 9 | 2015 | 68 |
AK314254.1 | - | 8 - 9 | 2306 | 68 | |
BC104901.1 | - | 8 - 9 | 1912 | 68 | |
BC104903.1 | - | 8 - 9 | 1906 | 68 |