The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102450.2 | NP_001095920.1 | 4 - 5 | 415 | 69 |
NM_033345.3 | NP_203131.1 | 3 - 4 | 286 | 69 | |
XM_005245555.3 | XP_005245612.1 | 4 - 5 | 429 | 69 | |
XM_011510089.2 | XP_011508391.1 | 4 - 5 | 1326 | 69 | |
XM_011510090.2 | XP_011508392.1 | 7 - 8 | 1156 | 69 | |
XM_011510091.2 | XP_011508393.1 | 3 - 4 | 303 | 69 | |
XM_017002631.1 | XP_016858120.1 | 7 - 8 | 776 | 69 | |
XM_017002632.1 | XP_016858121.1 | 5 - 6 | 607 | 69 | |
XM_017002633.1 | XP_016858122.1 | 4 - 5 | 805 | 69 | |
XM_017002634.1 | XP_016858123.1 | 4 - 5 | 484 | 69 | |
XM_017002635.1 | XP_016858124.1 | 5 - 6 | 931 | 69 | |
XM_017002636.1 | XP_016858125.1 | 3 - 4 | 413 | 69 | |
XM_017002637.1 | XP_016858126.1 | 4 - 5 | 611 | 69 | |
GenBank mRNA | AF300649.1 | - | 2 - 3 | 124 | 69 |
AK294660.1 | - | 4 - 5 | 382 | 69 | |
AK296514.1 | - | 4 - 5 | 375 | 69 | |
BC069677.1 | - | 4 - 5 | 364 | 69 | |
BC069718.1 | - | 3 - 4 | 236 | 69 | |
BC103865.1 | - | 4 - 5 | 391 | 69 | |
BC103866.1 | - | 4 - 5 | 391 | 69 | |
BC103867.1 | - | 4 - 5 | 391 | 69 | |
BX281622.1 | - | 3 - 4 | 297 | 69 | |
BX954018.1 | - | 3 - 4 | 190 | 69 |