The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018339.5 | NP_060809.3 | 2 - 3 | 591 | 132 |
GenBank mRNA | AK002011.1 | - | 2 - 3 | 568 | 132 |
AK315734.1 | - | 2 - 3 | 580 | 132 | |
BC007069.1 | - | 2 - 3 | 485 | 132 |