Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPYD
Entrez Gene ID
1806
Gene Name
dihydropyrimidine dehydrogenase
Gene Aliases
DHP, DHPDHASE, DPD
Chromosome Location
Chr.1: 97077743 - 97921059 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000110.3 NP_000101.2 18 - 19 2438 69
XM_005270562.3 XP_005270619.2 17 - 18 2222 69
XM_006710397.3 XP_006710460.1 18 - 19 2438 69
XM_017000507.1 XP_016855996.1 17 - 18 2321 69
XM_017000508.1 XP_016855997.1 20 - 21 2648 69
XM_017000509.1 XP_016855998.1 19 - 20 2546 69
XM_017000510.1 XP_016855999.1 18 - 19 2435 69
GenBank mRNA AB003063.1 - 18 - 19 2301 69
AK291217.1 - 18 - 19 2404 69
BC131777.1 - 18 - 19 2354 69
BC131778.1 - 18 - 19 2354 69
U09178.1 - 18 - 19 2382 69
U20938.1 - 18 - 19 2402 69

View Full Product Details