The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000110.3 | NP_000101.2 | 3 - 4 | 365 | 105 |
NM_001160301.1 | NP_001153773.1 | 3 - 4 | 365 | 105 | |
XM_005270562.3 | XP_005270619.2 | 3 - 4 | 365 | 105 | |
XM_006710397.3 | XP_006710460.1 | 3 - 4 | 365 | 105 | |
XM_017000507.1 | XP_016855996.1 | 2 - 3 | 248 | 105 | |
XM_017000508.1 | XP_016855997.1 | 3 - 4 | 360 | 105 | |
XM_017000509.1 | XP_016855998.1 | 3 - 4 | 360 | 105 | |
XM_017000510.1 | XP_016855999.1 | 2 - 3 | 249 | 105 | |
GenBank mRNA | AB003063.1 | - | 3 - 4 | 228 | 105 |
AK291217.1 | - | 3 - 4 | 331 | 105 | |
AK297515.1 | - | 2 - 3 | 192 | 105 | |
BC008379.1 | - | 3 - 4 | 252 | 105 | |
BC064027.1 | - | 3 - 4 | 291 | 105 | |
BC108742.1 | - | 3 - 4 | 324 | 105 | |
BC131777.1 | - | 3 - 4 | 281 | 105 | |
BC131778.1 | - | 3 - 4 | 281 | 105 | |
BT006740.1 | - | 3 - 4 | 228 | 105 | |
DA721123.1 | - | 3 - 4 | 365 | 105 | |
U09178.1 | - | 3 - 4 | 309 | 105 | |
U20938.1 | - | 3 - 4 | 329 | 105 |