The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001076683.1 | NP_001070151.1 | 9 - 10 | 1138 | 65 |
NM_001076684.2 | NP_001070152.1 | 9 - 10 | 1224 | 65 | |
NM_014233.3 | NP_055048.1 | 10 - 11 | 1335 | 65 | |
NR_045058.1 | 10 - 11 | 1532 | 65 | ||
XM_006722059.3 | XP_006722122.1 | 10 - 11 | 1374 | 65 | |
XM_006722060.2 | XP_006722123.1 | 9 - 10 | 1118 | 65 | |
XM_006722061.2 | XP_006722124.1 | 10 - 11 | 1202 | 65 | |
XM_017024995.1 | XP_016880484.1 | 10 - 11 | 1383 | 65 | |
XM_017024996.1 | XP_016880485.1 | 10 - 11 | 1335 | 65 | |
XM_017024997.1 | XP_016880486.1 | 10 - 11 | 1527 | 65 | |
XM_017024998.1 | XP_016880487.1 | 10 - 11 | 1243 | 65 | |
XM_017024999.1 | XP_016880488.1 | 9 - 10 | 1118 | 65 | |
XM_017025000.1 | XP_016880489.1 | 9 - 10 | 1176 | 65 | |
XM_017025001.1 | XP_016880490.1 | 9 - 10 | 1224 | 65 | |
XM_017025002.1 | XP_016880491.1 | 9 - 10 | 1104 | 65 | |
XM_017025003.1 | XP_016880492.1 | 9 - 10 | 1114 | 65 | |
XM_017025004.1 | XP_016880493.1 | 9 - 10 | 1106 | 65 | |
GenBank mRNA | AK291733.1 | - | 10 - 11 | 1160 | 65 |
AK292518.1 | - | 10 - 11 | 1532 | 65 | |
AK292577.1 | - | 10 - 11 | 1420 | 65 | |
AK296917.1 | - | 4 - 5 | 448 | 65 | |
AK298008.1 | - | 7 - 8 | 750 | 65 | |
AK308321.1 | - | 8 - 9 | 1201 | 65 | |
BC042297.1 | - | 9 - 10 | 1129 | 65 | |
X53390.1 | - | 9 - 10 | 1186 | 65 | |
X53461.1 | - | 9 - 10 | 1186 | 65 | |
X56687.1 | - | 9 - 10 | 1107 | 65 |