The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161561.2 | NP_001155033.1 | 20 - 21 | 2666 | 94 |
NM_001161563.2 | NP_001155035.1 | 20 - 21 | 2588 | 94 | |
NM_001161565.2 | NP_001155037.1 | 19 - 20 | 2501 | 94 | |
NM_015028.3 | NP_055843.1 | 21 - 22 | 2753 | 94 | |
GenBank mRNA | AB011123.2 | - | 21 - 22 | 2674 | 94 |
AF172264.1 | - | 21 - 22 | 2408 | 94 | |
AF172265.1 | - | 20 - 21 | 2321 | 94 | |
AF172266.1 | - | 20 - 21 | 2243 | 94 | |
AF172268.1 | - | 19 - 20 | 2156 | 94 | |
AK291056.1 | - | 19 - 20 | 2501 | 94 | |
BC150256.1 | - | 21 - 22 | 2674 | 94 |