The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161560.2 | NP_001155032.1 | 29 - 30 | 4022 | 144 |
NM_001161561.2 | NP_001155033.1 | 29 - 30 | 3959 | 144 | |
NM_001161562.2 | NP_001155034.1 | 28 - 29 | 3935 | 144 | |
NM_001161563.2 | NP_001155035.1 | 29 - 30 | 3881 | 144 | |
NM_001161564.2 | NP_001155036.1 | 28 - 29 | 3857 | 144 | |
NM_001161565.2 | NP_001155037.1 | 28 - 29 | 3794 | 144 | |
NM_001161566.2 | NP_001155038.1 | 27 - 28 | 3770 | 144 | |
NM_015028.3 | NP_055843.1 | 30 - 31 | 4046 | 144 | |
GenBank mRNA | AB011123.2 | - | 30 - 31 | 3967 | 144 |
AF172264.1 | - | 30 - 31 | 3701 | 144 | |
AF172265.1 | - | 29 - 30 | 3614 | 144 | |
AF172266.1 | - | 29 - 30 | 3536 | 144 | |
AF172267.1 | - | 29 - 30 | 3677 | 144 | |
AF172268.1 | - | 28 - 29 | 3449 | 144 | |
AF172269.1 | - | 28 - 29 | 3590 | 144 | |
AF172270.1 | - | 28 - 29 | 3512 | 144 | |
AF172271.1 | - | 27 - 28 | 3425 | 144 | |
AK026470.1 | - | 4 - 5 | 820 | 144 | |
AK291056.1 | - | 28 - 29 | 3794 | 144 | |
BC019081.2 | - | 9 - 10 | 1165 | 144 | |
BC055427.1 | - | 9 - 10 | 1734 | 144 | |
BC150256.1 | - | 30 - 31 | 3967 | 144 |