The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161560.2 | NP_001155032.1 | 5 - 6 | 763 | 72 |
NM_001161561.2 | NP_001155033.1 | 5 - 6 | 763 | 72 | |
NM_001161562.2 | NP_001155034.1 | 5 - 6 | 763 | 72 | |
NM_001161563.2 | NP_001155035.1 | 5 - 6 | 763 | 72 | |
NM_001161564.2 | NP_001155036.1 | 5 - 6 | 763 | 72 | |
NM_001161565.2 | NP_001155037.1 | 5 - 6 | 763 | 72 | |
NM_001161566.2 | NP_001155038.1 | 5 - 6 | 763 | 72 | |
NM_015028.3 | NP_055843.1 | 5 - 6 | 763 | 72 | |
GenBank mRNA | AB011123.2 | - | 5 - 6 | 684 | 72 |
AF172264.1 | - | 5 - 6 | 418 | 72 | |
AF172265.1 | - | 5 - 6 | 418 | 72 | |
AF172266.1 | - | 5 - 6 | 418 | 72 | |
AF172267.1 | - | 5 - 6 | 418 | 72 | |
AF172268.1 | - | 5 - 6 | 418 | 72 | |
AF172269.1 | - | 5 - 6 | 418 | 72 | |
AF172270.1 | - | 5 - 6 | 418 | 72 | |
AF172271.1 | - | 5 - 6 | 418 | 72 | |
AK291056.1 | - | 5 - 6 | 763 | 72 | |
BC150256.1 | - | 5 - 6 | 684 | 72 |