Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TINAGL1
Entrez Gene ID
64129
Gene Name
tubulointerstitial nephritis antigen like 1
Gene Aliases
ARG1, LCN7, LIECG3, TINAGRP
Chromosome Location
Chr.1: 31576384 - 31587686 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204414.1 NP_001191343.1 8 - 9 1126 60
NM_001204415.1 NP_001191344.1 8 - 9 827 60
NM_022164.2 NP_071447.1 9 - 10 1219 60
XM_005271107.2 XP_005271164.1 8 - 9 864 60
XM_011541946.1 XP_011540248.1 7 - 8 931 60
GenBank mRNA AB050716.1 - 9 - 10 1169 60
AF205436.1 - 9 - 10 1145 60
AF236150.1 - 9 - 10 1147 60
AK027839.1 - 3 - 4 363 60
AK075398.1 - 9 - 10 1192 60
AK292770.1 - 9 - 10 1193 60
AK298382.1 - 8 - 9 1096 60
AK301070.1 - 9 - 10 1229 60
AY358421.1 - 9 - 10 1192 60
BC009048.1 - 7 - 8 750 60
BC064633.1 - 9 - 10 1177 60

View Full Product Details