Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCL6
Entrez Gene ID
604
Gene Name
B-cell CLL/lymphoma 6
Gene Aliases
BCL5, BCL6A, LAZ3, ZBTB27, ZNF51
Chromosome Location
Chr.3: 187721377 - 187745725 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130845.1 NP_001124317.1 5 - 6 1813 85
NM_001134738.1 NP_001128210.1 4 - 5 1405 85
NM_001706.4 NP_001697.2 5 - 6 1721 85
XM_005247694.3 XP_005247751.1 5 - 6 5065 85
XM_011513062.2 XP_011511364.1 5 - 6 1721 85
GenBank mRNA AK304325.1 - 5 - 6 1464 85
AK308162.1 - 5 - 6 1463 85
BC142705.1 - 5 - 6 1436 85
BC150184.1 - 5 - 6 1437 85
BX649185.1 - 5 - 6 2343 85
EU139066.1 - 3 - 4 1380 85
EU883531.1 - 3 - 4 1356 85
S67779.1 - 5 - 6 1775 85
U00115.1 - 5 - 6 1683 85
Z21943.1 - 5 - 6 1440 85

View Full Product Details