The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005095.2 | NP_005086.2 | 12 - 13 | 2194 | 159 |
XM_005271328.4 | XP_005271385.1 | 12 - 13 | 2473 | 159 | |
XM_005271331.2 | XP_005271388.1 | 9 - 10 | 1439 | 159 | |
XM_011542421.2 | XP_011540723.1 | 12 - 13 | 2120 | 159 | |
XM_011542423.2 | XP_011540725.1 | 13 - 14 | 2525 | 159 | |
XM_011542425.2 | XP_011540727.1 | 10 - 11 | 1479 | 159 | |
XM_017002803.1 | XP_016858292.1 | 13 - 14 | 2247 | 159 | |
XM_017002804.1 | XP_016858293.1 | 13 - 14 | 2187 | 159 | |
XM_017002805.1 | XP_016858294.1 | 8 - 9 | 1685 | 159 | |
GenBank mRNA | AB007885.1 | - | 8 - 9 | 1314 | 159 |
AK309705.1 | - | 11 - 12 | 1854 | 159 | |
BC012093.1 | - | 10 - 11 | 1773 | 159 | |
BC127113.1 | - | 13 - 14 | 2200 | 159 | |
BC127114.1 | - | 9 - 10 | 1383 | 159 | |
BX647376.1 | - | 8 - 9 | 1424 | 159 | |
BX648067.1 | - | 3 - 4 | 488 | 159 | |
CR936741.1 | - | 11 - 12 | 2058 | 159 |