Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMYM4
Entrez Gene ID
9202
Gene Name
zinc finger MYM-type containing 4
Gene Aliases
CDIR, MYM, ZNF198L3, ZNF262
Chromosome Location
Chr.1: 35268694 - 35421944 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005095.2 NP_005086.2 20 - 21 3283 88
XM_005271328.4 XP_005271385.1 20 - 21 3562 88
XM_005271331.2 XP_005271388.1 17 - 18 2528 88
XM_011542421.2 XP_011540723.1 20 - 21 3209 88
XM_011542423.2 XP_011540725.1 21 - 22 3614 88
XM_011542425.2 XP_011540727.1 18 - 19 2568 88
XM_017002803.1 XP_016858292.1 21 - 22 3336 88
XM_017002804.1 XP_016858293.1 21 - 22 3276 88
XM_017002805.1 XP_016858294.1 16 - 17 2774 88
GenBank mRNA AB007885.1 - 16 - 17 2403 88
AK309705.1 - 19 - 20 2943 88
BC012093.1 - 18 - 19 2862 88
BC127113.1 - 21 - 22 3289 88
BC127114.1 - 17 - 18 2472 88
BX647376.1 - 16 - 17 2514 88
BX648067.1 - 11 - 12 1577 88
CR936741.1 - 19 - 20 3147 88

View Full Product Details