The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005095.2 | NP_005086.2 | 3 - 4 | 686 | 128 |
XM_005271328.4 | XP_005271385.1 | 3 - 4 | 965 | 128 | |
XM_011542421.2 | XP_011540723.1 | 3 - 4 | 612 | 128 | |
XM_011542423.2 | XP_011540725.1 | 4 - 5 | 1017 | 128 | |
XM_017002803.1 | XP_016858292.1 | 4 - 5 | 739 | 128 | |
XM_017002804.1 | XP_016858293.1 | 3 - 4 | 603 | 128 | |
XM_017002805.1 | XP_016858294.1 | 1 - 2 | 518 | 128 | |
GenBank mRNA | AK308320.1 | - | 3 - 4 | 686 | 128 |
BC012093.1 | - | 1 - 2 | 265 | 128 | |
BC127113.1 | - | 4 - 5 | 692 | 128 | |
CR936741.1 | - | 2 - 3 | 550 | 128 |