Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSF3R
Entrez Gene ID
1441
Gene Name
colony stimulating factor 3 receptor
Gene Aliases
CD114, GCSFR, SCN7
Chromosome Location
Chr.1: 36466043 - 36483314 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000760.3 NP_000751.1 14 - 15 2454 60
NM_156039.3 NP_724781.1 14 - 15 2454 60
NM_172313.2 NP_758519.1 14 - 15 2454 60
XM_005270493.1 XP_005270550.1 14 - 15 2454 60
XM_011540748.2 XP_011539050.1 13 - 14 1979 60
XM_011540749.1 XP_011539051.1 14 - 15 2454 60
XM_011540750.1 XP_011539052.1 9 - 10 1284 60
XM_017000370.1 XP_016855859.1 13 - 14 2016 60
GenBank mRNA AB209656.1 - 7 - 8 4718 60
BC053585.1 - 14 - 15 2047 60
DQ424902.1 - 12 - 13 1870 60
M59818.1 - 14 - 15 2039 60
M59820.1 - 14 - 15 2039 60
X55720.1 - 14 - 15 2034 60
X55721.1 - 14 - 15 2035 60

View Full Product Details