The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005082.4 | NP_005073.2 | 4 - 5 | 1149 | 80 |
GenBank mRNA | AB208994.1 | - | 4 - 5 | 1133 | 80 |
AK310720.1 | - | 5 - 6 | 634 | 80 | |
BC016924.1 | - | 4 - 5 | 1133 | 80 | |
BC042541.1 | - | 4 - 5 | 1133 | 80 | |
CF121587.1 | - | 1 - 2 | 116 | 80 | |
D21205.1 | - | 4 - 5 | 1128 | 80 |