Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLSCR2
Entrez Gene ID
57047
Gene Name
phospholipid scramblase 2
Gene Aliases
-
Chromosome Location
Chr.3: 146391363 - 146497163 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011513013.2 XP_011511315.2 8 - 9 4783 80
XM_017006898.1 XP_016862387.1 9 - 10 3022 80
XM_017006901.1 XP_016862390.1 9 - 10 4656 80
XM_017006905.1 XP_016862394.1 10 - 11 4239 80
XM_017006906.1 XP_016862395.1 10 - 11 4020 80
XM_017006907.1 XP_016862396.1 10 - 11 3785 80
XM_017006908.1 XP_016862397.1 10 - 11 3910 80
XM_017006909.1 XP_016862398.1 9 - 10 3697 80
XM_017006912.1 XP_016862401.1 8 - 9 3438 80
XM_017006914.1 XP_016862403.1 8 - 9 3645 80
XM_017006915.1 XP_016862404.1 8 - 9 3627 80

View Full Product Details