The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282951.2 | NP_001269880.1 | 1 - 2 | 220 | 65 |
XM_006712693.1 | XP_006712756.1 | 1 - 2 | 220 | 65 | |
XM_011511640.1 | XP_011509942.1 | 1 - 2 | 197 | 65 | |
XM_017004716.1 | XP_016860205.1 | 1 - 2 | 220 | 65 | |
XM_017004717.1 | XP_016860206.1 | 1 - 2 | 197 | 65 | |
GenBank mRNA | BI906506.1 | - | 1 - 2 | 149 | 65 |
DA754035.1 | - | 1 - 2 | 221 | 65 |