The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319224.1 | NP_001306153.1 | 2 - 3 | 461 | 83 |
NM_003686.4 | NP_003677.4 | 1 - 2 | 298 | 83 | |
NM_006027.4 | NP_006018.4 | 1 - 2 | 298 | 83 | |
XM_011544321.1 | XP_011542623.1 | 2 - 3 | 463 | 83 | |
XM_011544322.1 | XP_011542624.1 | 2 - 3 | 532 | 83 | |
XM_017002793.1 | XP_016858282.1 | 2 - 3 | 463 | 83 | |
GenBank mRNA | AF042282.1 | - | 1 - 2 | 202 | 83 |
AF060479.1 | - | 1 - 2 | 193 | 83 | |
AF084974.1 | - | 1 - 2 | 193 | 83 | |
AF091740.1 | - | 1 - 2 | 195 | 83 | |
AK291291.1 | - | 2 - 3 | 449 | 83 | |
BM465399.1 | - | - | 463 | 83 | |
CD644038.1 | - | 2 - 3 | 429 | 83 | |
DA479425.1 | - | 2 - 3 | 464 | 83 |