Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHACTR1
Entrez Gene ID
221692
Gene Name
phosphatase and actin regulator 1
Gene Aliases
RPEL, RPEL1, dJ257A7.2
Chromosome Location
Chr.6: 12716635 - 13290484 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242648.2 NP_001229577.1 12 - 13 1845 72
NM_001322308.1 NP_001309237.1 11 - 12 1800 72
NM_001322309.1 NP_001309238.1 11 - 12 1691 72
NM_001322310.1 NP_001309239.1 11 - 12 1766 72
NM_001322311.1 NP_001309240.1 9 - 10 1452 72
NM_001322312.1 NP_001309241.1 9 - 10 1815 72
NM_001322313.1 NP_001309242.1 10 - 11 2022 72
NM_001322314.1 NP_001309243.1 10 - 11 1954 72
NM_030948.3 NP_112210.1 12 - 13 2035 72
XM_005248934.2 XP_005248991.1 13 - 14 2084 72
XM_017010452.1 XP_016865941.1 10 - 11 1748 72
XM_017010454.1 XP_016865943.1 13 - 14 2084 72
XM_017010455.1 XP_016865944.1 13 - 14 2052 72
XM_017010456.1 XP_016865945.1 12 - 13 2049 72
XM_017010457.1 XP_016865946.1 12 - 13 2028 72
XM_017010458.1 XP_016865947.1 9 - 10 1541 72
XM_017010459.1 XP_016865948.1 12 - 13 1877 72
XM_017010460.1 XP_016865949.1 12 - 13 1845 72
XM_017010461.1 XP_016865950.1 11 - 12 1816 72
XM_017010462.1 XP_016865951.1 11 - 12 1842 72
XM_017010463.1 XP_016865952.1 10 - 11 1942 72
XM_017010464.1 XP_016865953.1 9 - 10 1541 72
XM_017010465.1 XP_016865954.1 10 - 11 1722 72
XM_017010466.1 XP_016865955.1 10 - 11 1735 72
XM_017010467.1 XP_016865956.1 9 - 10 1722 72
XM_017010469.1 XP_016865958.1 9 - 10 1524 72
GenBank mRNA AK090769.1 - 11 - 12 1702 72
AK290017.1 - 12 - 13 1696 72
BC101526.1 - 11 - 12 1609 72
BC101528.1 - 11 - 12 1609 72
BF026507.1 - - 216 72
BQ434462.1 - 2 - 3 252 72

View Full Product Details