The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016467.4 | NP_057551.1 | 1 - 2 | 103 | 112 |
GenBank mRNA | AK313533.1 | - | 1 - 2 | 67 | 112 |