Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RALGPS1
Entrez Gene ID
9649
Gene Name
Ral GEF with PH domain and SH3 binding motif 1
Gene Aliases
RALGEF2, RALGPS1A
Chromosome Location
Chr.9: 126914774 - 127223166 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190728.1 NP_001177657.1 14 - 15 1381 76
NM_001322322.1 NP_001309251.1 17 - 18 1827 76
XM_011519226.1 XP_011517528.1 17 - 18 1753 76
XM_017015346.1 XP_016870835.1 15 - 16 1507 76
GenBank mRNA AF221098.1 - 14 - 15 1381 76

View Full Product Details