Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM37
Entrez Gene ID
4591
Gene Name
tripartite motif containing 37
Gene Aliases
MUL, POB1, TEF3
Chromosome Location
Chr.17: 58968068 - 59106921 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005207.3 NP_001005207.1 10 - 11 1320 78
NM_001320987.1 NP_001307916.1 9 - 10 1218 78
NM_001320988.1 NP_001307917.1 10 - 11 1320 78
NM_001320989.1 NP_001307918.1 10 - 11 1320 78
NM_001320990.1 NP_001307919.1 11 - 12 1396 78
NM_015294.4 NP_056109.1 10 - 11 1320 78
XM_005257385.1 XP_005257442.1 10 - 11 1152 78
XM_011524831.1 XP_011523133.1 10 - 11 1152 78
XM_011524832.2 XP_011523134.1 10 - 11 1152 78
XM_011524834.1 XP_011523136.1 9 - 10 1034 78
XM_011524836.1 XP_011523138.1 10 - 11 1152 78
XM_017024662.1 XP_016880151.1 10 - 11 1152 78
XM_017024663.1 XP_016880152.1 10 - 11 1152 78
XM_017024665.1 XP_016880154.1 9 - 10 1034 78
XM_017024668.1 XP_016880157.1 9 - 10 1034 78
XM_017024670.1 XP_016880159.1 9 - 10 1033 78
XM_017024671.1 XP_016880160.1 9 - 10 1064 78
XM_017024672.1 XP_016880161.1 9 - 10 1064 78
XM_017024673.1 XP_016880162.1 9 - 10 856 78
XM_017024674.1 XP_016880163.1 9 - 10 856 78
GenBank mRNA AB020705.1 - 10 - 11 905 78
AF213365.1 - 10 - 11 1124 78
AK025648.1 - 4 - 5 338 78
AK289674.1 - 10 - 11 1135 78
AK292459.1 - 10 - 11 1192 78
AK294850.1 - 9 - 10 1123 78
BC036012.1 - 10 - 11 1156 78
BX537955.1 - 11 - 12 1215 78

View Full Product Details