Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM37
Entrez Gene ID
4591
Gene Name
tripartite motif containing 37
Gene Aliases
MUL, POB1, TEF3
Chromosome Location
Chr.17: 58968068 - 59106921 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005207.3 NP_001005207.1 12 - 13 1468 80
NM_001320987.1 NP_001307916.1 11 - 12 1366 80
NM_001320988.1 NP_001307917.1 12 - 13 1468 80
NM_001320989.1 NP_001307918.1 12 - 13 1468 80
NM_001320990.1 NP_001307919.1 13 - 14 1544 80
NM_015294.4 NP_056109.1 12 - 13 1468 80
XM_005257385.1 XP_005257442.1 12 - 13 1300 80
XM_011524831.1 XP_011523133.1 12 - 13 1300 80
XM_011524832.2 XP_011523134.1 12 - 13 1300 80
XM_011524833.1 XP_011523135.1 11 - 12 1249 80
XM_011524834.1 XP_011523136.1 11 - 12 1182 80
XM_011524836.1 XP_011523138.1 12 - 13 1300 80
XM_017024662.1 XP_016880151.1 12 - 13 1300 80
XM_017024663.1 XP_016880152.1 12 - 13 1300 80
XM_017024664.1 XP_016880153.1 11 - 12 1249 80
XM_017024665.1 XP_016880154.1 11 - 12 1182 80
XM_017024666.1 XP_016880155.1 11 - 12 1249 80
XM_017024667.1 XP_016880156.1 11 - 12 1249 80
XM_017024668.1 XP_016880157.1 11 - 12 1182 80
XM_017024669.1 XP_016880158.1 10 - 11 1131 80
XM_017024670.1 XP_016880159.1 11 - 12 1181 80
XM_017024671.1 XP_016880160.1 11 - 12 1212 80
XM_017024672.1 XP_016880161.1 11 - 12 1212 80
XM_017024673.1 XP_016880162.1 11 - 12 1004 80
XM_017024674.1 XP_016880163.1 11 - 12 1004 80
GenBank mRNA AB020705.1 - 12 - 13 1053 80
AF213365.1 - 12 - 13 1272 80
AK025648.1 - 6 - 7 486 80
AK289674.1 - 12 - 13 1283 80
AK292459.1 - 12 - 13 1340 80
AK294850.1 - 11 - 12 1271 80
BC036012.1 - 12 - 13 1304 80
BX537955.1 - 13 - 14 1363 80

View Full Product Details