The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000382.2 | NP_000373.1 | 3 - 4 | 688 | 139 |
NM_001031806.1 | NP_001026976.1 | 3 - 4 | 688 | 139 | |
XM_011523732.1 | XP_011522034.1 | 4 - 5 | 597 | 139 | |
XM_011523733.1 | XP_011522035.1 | 4 - 5 | 634 | 139 | |
XM_017024355.1 | XP_016879844.1 | 3 - 4 | 490 | 139 | |
XM_017024356.1 | XP_016879845.1 | 4 - 5 | 596 | 139 | |
XM_017024357.1 | XP_016879846.1 | 4 - 5 | 597 | 139 | |
XM_017024358.1 | XP_016879847.1 | 4 - 5 | 634 | 139 | |
GenBank mRNA | AB208894.1 | - | 3 - 4 | 396 | 139 |
AK000858.1 | - | 2 - 3 | 298 | 139 | |
AK292381.1 | - | 3 - 4 | 655 | 139 | |
AK315096.1 | - | 3 - 4 | 517 | 139 | |
BC002430.2 | - | 3 - 4 | 508 | 139 | |
CR457422.1 | - | 3 - 4 | 467 | 139 | |
L47162.1 | - | 3 - 4 | 630 | 139 | |
U46689.1 | - | 3 - 4 | 916 | 139 |