The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014877.3 | NP_055692.2 | 26 - 27 | 3811 | 76 |
XM_005257888.4 | XP_005257945.1 | 27 - 28 | 3940 | 76 | |
XM_005257889.4 | XP_005257946.1 | 26 - 27 | 3862 | 76 | |
XM_006722214.3 | XP_006722277.1 | 27 - 28 | 3943 | 76 | |
XM_006722215.3 | XP_006722278.1 | 25 - 26 | 3815 | 76 | |
XM_006722216.3 | XP_006722279.1 | 16 - 17 | 2645 | 76 | |
XM_011525544.2 | XP_011523846.1 | 27 - 28 | 4014 | 76 | |
XM_017025477.1 | XP_016880966.1 | 24 - 25 | 3731 | 76 | |
XM_017025478.1 | XP_016880967.1 | 27 - 28 | 3996 | 76 | |
GenBank mRNA | BC094881.1 | - | 24 - 25 | 3837 | 76 |
BC130582.1 | - | 26 - 27 | 3791 | 76 | |
BC144083.1 | - | 26 - 27 | 3794 | 76 | |
D29677.2 | - | 25 - 26 | 3769 | 76 |