The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014877.3 | NP_055692.2 | 28 - 29 | 4102 | 150 |
XM_005257888.4 | XP_005257945.1 | 29 - 30 | 4231 | 150 | |
XM_005257889.4 | XP_005257946.1 | 28 - 29 | 4153 | 150 | |
XM_006722214.3 | XP_006722277.1 | 29 - 30 | 4234 | 150 | |
XM_006722215.3 | XP_006722278.1 | 27 - 28 | 4106 | 150 | |
XM_006722216.3 | XP_006722279.1 | 18 - 19 | 2936 | 150 | |
XM_011525544.2 | XP_011523846.1 | 29 - 30 | 4305 | 150 | |
XM_017025477.1 | XP_016880966.1 | 26 - 27 | 4022 | 150 | |
XM_017025478.1 | XP_016880967.1 | 29 - 30 | 4287 | 150 | |
GenBank mRNA | BC094881.1 | - | 26 - 27 | 4128 | 150 |
BC130582.1 | - | 28 - 29 | 4082 | 150 | |
BC144083.1 | - | 28 - 29 | 4085 | 150 | |
D29677.2 | - | 27 - 28 | 4060 | 150 |