The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168215.1 | NP_001161687.1 | 4 - 5 | 279 | 66 |
NM_153229.2 | NP_694961.2 | 3 - 4 | 244 | 66 | |
XM_006721730.3 | XP_006721793.1 | 3 - 4 | 244 | 66 | |
GenBank mRNA | AK090637.1 | - | 3 - 4 | 199 | 66 |
AY358204.1 | - | 4 - 5 | 279 | 66 | |
BC064507.1 | - | 3 - 4 | 172 | 66 | |
DB072318.1 | - | 3 - 4 | 245 | 66 |