Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFSF8
Entrez Gene ID
944
Gene Name
tumor necrosis factor superfamily member 8
Gene Aliases
CD153, CD30L, CD30LG, TNLG3A
Chromosome Location
Chr.9: 114893343 - 114930595 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244.3 NP_001235.1 2 - 3 528 100
NM_001252290.1 NP_001239219.1 2 - 3 528 100
GenBank mRNA AK313619.1 - 2 - 3 528 100
BC093630.1 - 2 - 3 310 100
BC111939.1 - 2 - 3 310 100
CD690231.1 - 2 - 3 238 100
L09753.1 - 2 - 3 350 100

View Full Product Details