The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168319.1 | NP_001161791.1 | 3 - 4 | 721 | 134 |
NM_001955.4 | NP_001946.3 | 3 - 4 | 724 | 134 | |
XM_011514330.2 | XP_011512632.1 | 4 - 5 | 480 | 134 | |
XM_011514331.2 | XP_011512633.1 | 5 - 6 | 641 | 134 | |
XM_011514332.2 | XP_011512634.1 | 4 - 5 | 478 | 134 | |
XM_017010331.1 | XP_016865820.1 | 4 - 5 | 664 | 134 | |
GenBank mRNA | AK226096.1 | - | 3 - 4 | 639 | 134 |
AK291838.1 | - | 3 - 4 | 661 | 134 | |
AK313914.1 | - | 3 - 4 | 656 | 134 | |
BC009720.1 | - | 3 - 4 | 593 | 134 | |
BX439515.2 | - | 3 - 4 | 684 | 134 | |
CR541903.1 | - | 3 - 4 | 390 | 134 | |
S56805.1 | - | 3 - 4 | 726 | 134 | |
Y00749.1 | - | 3 - 4 | 642 | 134 |