Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS9D1
Entrez Gene ID
9605
Gene Name
VPS9 domain containing 1
Gene Aliases
ATP-BL, C16orf7
Chromosome Location
Chr.16: 89707133 - 89721046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004913.2 NP_004904.2 4 - 5 549 109
XM_005256329.4 XP_005256386.1 4 - 5 609 109
XM_005256330.4 XP_005256387.1 4 - 5 426 109
XM_006721350.2 XP_006721413.1 4 - 5 609 109
XM_011523476.2 XP_011521778.1 4 - 5 606 109
XM_011523477.2 XP_011521779.1 3 - 4 2422 109
XM_011523478.2 XP_011521780.1 3 - 4 2419 109
XM_011523479.2 XP_011521781.1 4 - 5 609 109
XM_011523480.2 XP_011521782.1 4 - 5 463 109
GenBank mRNA AB018551.1 - 3 - 4 590 109
BY795348.2 - 1 - 2 170 109

View Full Product Details