The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004913.2 | NP_004904.2 | 9 - 10 | 962 | 145 |
XM_005256329.4 | XP_005256386.1 | 9 - 10 | 1022 | 145 | |
XM_005256330.4 | XP_005256387.1 | 9 - 10 | 839 | 145 | |
XM_005256331.2 | XP_005256388.1 | 6 - 7 | 496 | 145 | |
XM_006721350.2 | XP_006721413.1 | 9 - 10 | 1022 | 145 | |
XM_011523476.2 | XP_011521778.1 | 9 - 10 | 1019 | 145 | |
XM_011523477.2 | XP_011521779.1 | 8 - 9 | 2835 | 145 | |
XM_011523478.2 | XP_011521780.1 | 8 - 9 | 2832 | 145 | |
XM_011523479.2 | XP_011521781.1 | 9 - 10 | 1022 | 145 | |
XM_011523480.2 | XP_011521782.1 | 9 - 10 | 876 | 145 | |
GenBank mRNA | AB018551.1 | - | 8 - 9 | 1003 | 145 |