The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128141.2 | NP_001121613.1 | 4 - 5 | 572 | 78 |
NM_004413.3 | NP_004404.1 | 4 - 5 | 660 | 78 | |
XM_005256285.4 | XP_005256342.1 | 4 - 5 | 610 | 78 | |
XM_005256286.3 | XP_005256343.1 | 4 - 5 | 522 | 78 | |
XM_011522925.2 | XP_011521227.1 | 4 - 5 | 620 | 78 | |
XM_017023009.1 | XP_016878498.1 | 4 - 5 | 683 | 78 | |
GenBank mRNA | BC017023.1 | - | 3 - 4 | 491 | 78 |
BT006664.1 | - | 3 - 4 | 369 | 78 | |
CR457404.1 | - | 3 - 4 | 369 | 78 | |
D13138.1 | - | 3 - 4 | 412 | 78 | |
J05257.1 | - | 4 - 5 | 664 | 78 |