Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMD3
Entrez Gene ID
257019
Gene Name
FERM domain containing 3
Gene Aliases
4.1O, EPB41L4O, EPB41LO, P410
Chromosome Location
Chr.9: 83242990 - 83560208 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244959.1 NP_001231888.1 13 - 14 1394 107
NM_001244960.1 NP_001231889.1 13 - 14 1170 107
NM_001244961.1 NP_001231890.1 8 - 9 706 107
NM_174938.5 NP_777598.3 13 - 14 1394 107
XM_017014588.1 XP_016870077.1 13 - 14 1084 107
XM_017014589.1 XP_016870078.1 8 - 9 3521 107
XM_017014590.1 XP_016870079.1 12 - 13 1307 107
GenBank mRNA AK094281.1 - 13 - 14 1095 107
AK223597.1 - 13 - 14 1321 107
AK297722.1 - 13 - 14 1312 107
AY137774.1 - 13 - 14 1178 107
BC037253.1 - 8 - 9 706 107
EF560742.1 - 13 - 14 1245 107

View Full Product Details