The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281303.1 | NP_001268232.1 | 11 - 12 | 1179 | 96 |
NM_006415.3 | NP_006406.1 | 11 - 12 | 1179 | 96 | |
XM_011518139.2 | XP_011516441.1 | 11 - 12 | 1163 | 96 | |
XM_017014200.1 | XP_016869689.1 | 12 - 13 | 1224 | 96 | |
XM_017014201.1 | XP_016869690.1 | 12 - 13 | 1296 | 96 | |
XM_017014202.1 | XP_016869691.1 | 7 - 8 | 1357 | 96 | |
GenBank mRNA | AK291546.1 | - | 11 - 12 | 1079 | 96 |
AK299153.1 | - | 11 - 12 | 994 | 96 | |
AK309775.1 | - | - | 951 | 96 | |
BC068537.1 | - | 11 - 12 | 1083 | 96 | |
Y08685.1 | - | 11 - 12 | 1075 | 96 |