The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002389.4 | NP_002380.3 | 2 - 3 | 460 | 78 |
NM_153826.3 | NP_722548.1 | 2 - 3 | 460 | 78 | |
NM_172350.2 | NP_758860.1 | 2 - 3 | 460 | 78 | |
NM_172351.2 | NP_758861.1 | 2 - 3 | 460 | 78 | |
NM_172352.2 | NP_758862.1 | 2 - 3 | 460 | 78 | |
NM_172353.2 | NP_758863.1 | 2 - 3 | 460 | 78 | |
NM_172359.2 | NP_758869.1 | 2 - 3 | 460 | 78 | |
NM_172361.2 | NP_758871.1 | 2 - 3 | 460 | 78 | |
XM_011509563.1 | XP_011507865.1 | 2 - 3 | 460 | 78 | |
XM_011509564.1 | XP_011507866.1 | 2 - 3 | 460 | 78 | |
XM_017001308.1 | XP_016856797.1 | 2 - 3 | 460 | 78 | |
XM_017001309.1 | XP_016856798.1 | 2 - 3 | 460 | 78 | |
XM_017001310.1 | XP_016856799.1 | 2 - 3 | 460 | 78 | |
GenBank mRNA | A18585.1 | - | 2 - 3 | 318 | 78 |
AF209712.1 | - | 2 - 3 | 285 | 78 | |
AF209713.1 | - | 2 - 3 | 285 | 78 | |
AF209714.1 | - | 2 - 3 | 285 | 78 | |
AK222822.1 | - | 2 - 3 | 386 | 78 | |
AK296819.1 | - | 2 - 3 | 412 | 78 | |
AU131238.2 | - | 2 - 3 | 441 | 78 | |
BC030594.2 | - | 2 - 3 | 400 | 78 | |
BX640613.1 | - | 2 - 3 | 442 | 78 | |
BX649050.1 | - | 2 - 3 | 431 | 78 | |
D84105.1 | - | 2 - 3 | 306 | 78 | |
DB078198.1 | - | 2 - 3 | 460 | 78 | |
EF076055.1 | - | 2 - 3 | 318 | 78 | |
EF076056.1 | - | 2 - 3 | 318 | 78 | |
EF076057.1 | - | 2 - 3 | 318 | 78 | |
EF076058.1 | - | 2 - 3 | 318 | 78 | |
M58050.1 | - | 2 - 3 | 319 | 78 | |
S51940.1 | - | 2 - 3 | 372 | 78 | |
X59405.1 | - | 2 - 3 | 430 | 78 | |
X59406.1 | - | 2 - 3 | 389 | 78 | |
X59407.1 | - | 2 - 3 | 389 | 78 | |
X59408.1 | - | 2 - 3 | 430 | 78 | |
X59409.1 | - | 2 - 3 | 389 | 78 | |
X59410.1 | - | 2 - 3 | 389 | 78 | |
Y00651.1 | - | 2 - 3 | 328 | 78 |