Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CENPI
Entrez Gene ID
2491
Gene Name
centromere protein I
Gene Aliases
CENP-I, FSHPRH1, LRPR1
Chromosome Location
Chr.X: 101098159 - 101181847 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318521.1 NP_001305450.1 9 - 10 948 81
NM_001318523.1 NP_001305452.1 7 - 8 785 81
NM_006733.3 NP_006724.2 8 - 9 1042 81
XM_005262111.2 XP_005262168.1 9 - 10 912 81
XM_011530894.2 XP_011529196.1 9 - 10 941 81
XM_011530895.2 XP_011529197.1 10 - 11 1228 81
XM_011530897.2 XP_011529199.1 10 - 11 1103 81
XM_011530899.2 XP_011529201.1 8 - 9 909 81
XM_017029383.1 XP_016884872.1 9 - 10 941 81
GenBank mRNA AK302986.1 - 9 - 10 941 81
BC012462.2 - 7 - 8 786 81

View Full Product Details