The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014229.2 | NP_055044.1 | 7 - 8 | 1057 | 72 |
XM_011534033.2 | XP_011532335.1 | 7 - 8 | 1057 | 72 | |
XM_017007073.1 | XP_016862562.1 | 8 - 9 | 908 | 72 | |
GenBank mRNA | AK312719.1 | - | 7 - 8 | 1057 | 72 |
S75989.1 | - | 7 - 8 | 1025 | 72 |