Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D5
Entrez Gene ID
9779
Gene Name
TBC1 domain family member 5
Gene Aliases
-
Chromosome Location
Chr.3: 17157162 - 17742739 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134381.1 NP_001127853.1 11 - 12 971 72
NM_014744.2 NP_055559.1 10 - 11 2372 72
XM_005265611.1 XP_005265668.1 10 - 11 942 72
XM_005265612.1 XP_005265669.1 9 - 10 877 72
XM_005265614.1 XP_005265671.1 10 - 11 942 72
XM_005265615.1 XP_005265672.1 9 - 10 872 72
XM_005265616.4 XP_005265673.1 8 - 9 801 72
XM_006713430.1 XP_006713493.1 10 - 11 992 72
XM_011534281.1 XP_011532583.1 12 - 13 1253 72
XM_011534283.2 XP_011532585.1 11 - 12 1031 72
XM_011534284.2 XP_011532586.1 11 - 12 1076 72
XM_011534286.1 XP_011532588.1 10 - 11 1008 72
XM_011534287.1 XP_011532589.1 10 - 11 990 72
XM_017007552.1 XP_016863041.1 13 - 14 1354 72
XM_017007553.1 XP_016863042.1 13 - 14 1384 72
XM_017007554.1 XP_016863043.1 18 - 19 1997 72
XM_017007555.1 XP_016863044.1 12 - 13 1236 72
XM_017007556.1 XP_016863045.1 11 - 12 1216 72
XM_017007557.1 XP_016863046.1 9 - 10 836 72
XM_017007558.1 XP_016863047.1 9 - 10 866 72
XM_017007559.1 XP_016863048.1 11 - 12 1216 72
XM_017007560.1 XP_016863049.1 11 - 12 1046 72
XM_017007561.1 XP_016863050.1 9 - 10 877 72
XM_017007562.1 XP_016863051.1 10 - 11 990 72
XM_017007563.1 XP_016863052.1 8 - 9 796 72
XM_017007564.1 XP_016863053.1 9 - 10 872 72
XM_017007565.1 XP_016863054.1 10 - 11 1170 72
XM_017007566.1 XP_016863055.1 10 - 11 959 72
XM_017007567.1 XP_016863056.1 8 - 9 800 72
XM_017007568.1 XP_016863057.1 9 - 10 836 72
GenBank mRNA AB449886.1 - 8 - 9 707 72
AK309374.1 - 6 - 7 632 72
AK310539.1 - 11 - 12 971 72
BC013145.1 - 9 - 10 884 72
D86965.1 - 10 - 11 2371 72

View Full Product Details