The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020857.2 | NP_065908.1 | 1 - 2 | 433 | 102 |
XM_011521844.2 | XP_011520146.1 | 1 - 2 | 452 | 102 | |
XM_017022447.1 | XP_016877936.1 | 1 - 2 | 453 | 102 | |
GenBank mRNA | AB040908.1 | - | 1 - 2 | 408 | 102 |
AF308802.1 | - | 1 - 2 | 408 | 102 | |
AL713725.1 | - | 1 - 2 | 433 | 102 | |
BC001513.2 | - | 1 - 2 | 260 | 102 |