The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005207.3 | NP_001005207.1 | 24 - 25 | 3342 | 84 |
NM_001320989.1 | NP_001307918.1 | 23 - 24 | 3225 | 84 | |
XM_005257385.1 | XP_005257442.1 | 24 - 25 | 3201 | 84 | |
XM_017024667.1 | XP_016880156.1 | 23 - 24 | 3123 | 84 | |
GenBank mRNA | AI307801.1 | - | 1 - 2 | 83 | |
AK292459.1 | - | 24 - 25 | 3214 | 84 | |
BC036012.1 | - | 24 - 25 | 3178 | 84 | |
BQ447834.1 | - | 1 - 2 | 84 |