The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322250.1 | NP_001309179.1 | 5 - 6 | 385 | 85 |
NM_001322251.1 | NP_001309180.1 | 5 - 6 | 489 | 85 | |
NM_138962.3 | NP_620412.1 | 5 - 6 | 489 | 85 | |
NM_170721.1 | NP_733839.1 | 4 - 5 | 339 | 85 | |
XM_005257014.3 | XP_005257071.1 | 5 - 6 | 414 | 85 | |
XM_005257015.3 | XP_005257072.1 | 5 - 6 | 414 | 85 | |
XM_017024148.1 | XP_016879637.1 | 5 - 6 | 414 | 85 | |
GenBank mRNA | AK093888.1 | - | 4 - 5 | 339 | 85 |
AK295060.1 | - | 5 - 6 | 386 | 85 |