The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024308.3 | NP_077284.2 | 2 - 3 | 578 | 62 |
XM_005257658.2 | XP_005257715.1 | 2 - 3 | 583 | 62 | |
GenBank mRNA | AK026196.1 | - | 2 - 3 | 369 | 62 |
AK315735.1 | - | 2 - 3 | 499 | 62 | |
AY358712.1 | - | 2 - 3 | 475 | 62 | |
BC002731.2 | - | 2 - 3 | 464 | 62 | |
CR457356.1 | - | 1 - 2 | 127 | 62 | |
LC110386.1 | - | 2 - 3 | 364 | 62 |