Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COBLL1
Entrez Gene ID
22837
Gene Name
cordon-bleu WH2 repeat protein like 1
Gene Aliases
COBLR1
Chromosome Location
Chr.2: 164680185 - 164842168 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278458.1 NP_001265387.1 12 - 13 1664 91
NM_014900.4 NP_055715.3 9 - 10 1564 91
GenBank mRNA AB023194.1 - 9 - 10 1558 91
AK002054.1 - 12 - 13 1589 91
AK294937.1 - 12 - 13 1664 91

View Full Product Details