The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000814.5 | NP_000805.1 | 6 - 7 | 798 | 64 |
NM_001191320.1 | NP_001178249.1 | 4 - 5 | 596 | 64 | |
NM_001191321.2 | NP_001178250.1 | 4 - 5 | 646 | 64 | |
NM_001278631.1 | NP_001265560.1 | 7 - 8 | 894 | 64 | |
NM_021912.4 | NP_068712.1 | 6 - 7 | 748 | 64 | |
XM_011521428.2 | XP_011519730.1 | 5 - 6 | 781 | 64 | |
GenBank mRNA | AK295167.1 | - | 4 - 5 | 562 | 64 |
AK302822.1 | - | 4 - 5 | 596 | 64 | |
AK315311.1 | - | 6 - 7 | 749 | 64 | |
BC010641.1 | - | 6 - 7 | 764 | 64 | |
CD013894.1 | - | 7 - 8 | 833 | 64 | |
CD652458.1 | - | 6 - 7 | 649 | 64 | |
KJ534842.1 | - | 6 - 7 | 684 | 64 | |
M82919.1 | - | 6 - 7 | 737 | 64 |