The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242837.1 | NP_001229766.1 | 19 - 20 | 2757 | 82 |
NM_012305.3 | NP_036437.1 | 19 - 20 | 2754 | 82 | |
XM_011519929.1 | XP_011518231.1 | 19 - 20 | 2514 | 82 | |
GenBank mRNA | AB020706.2 | - | 19 - 20 | 2541 | 82 |
AK027640.1 | - | 11 - 12 | 1491 | 82 | |
AK027891.1 | - | 18 - 19 | 2499 | 82 | |
AK095725.1 | - | 1 - 2 | 2512 | 82 | |
AK223558.1 | - | 19 - 20 | 2685 | 82 | |
AK293763.1 | - | 14 - 15 | 1830 | 82 | |
AK299364.1 | - | 19 - 20 | 2532 | 82 | |
AL117614.1 | - | 6 - 7 | 645 | 82 | |
BC006155.1 | - | 19 - 20 | 2682 | 82 |