The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145418.1 | NP_001138890.1 | 22 - 23 | 5954 | 95 |
XM_005261405.2 | XP_005261462.1 | 21 - 22 | 5930 | 95 | |
XM_006724171.3 | XP_006724234.1 | 21 - 22 | 14732 | 95 | |
XM_011530018.2 | XP_011528320.1 | 22 - 23 | 6133 | 95 | |
XM_017028673.1 | XP_016884162.1 | 22 - 23 | 5927 | 95 | |
GenBank mRNA | AB028966.3 | - | 17 - 18 | 4747 | 95 |
AB665284.1 | - | 22 - 23 | 5902 | 95 | |
AL080174.1 | - | 1 - 2 | 85 | 95 | |
BC016465.1 | - | 14 - 15 | 2498 | 95 |