Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC28
Entrez Gene ID
23331
Gene Name
tetratricopeptide repeat domain 28
Gene Aliases
TPRBK
Chromosome Location
Chr.22: 27978014 - 28679865 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145418.1 NP_001138890.1 19 - 20 5691 93
XM_006724171.3 XP_006724234.1 18 - 19 14469 93
XM_011530018.2 XP_011528320.1 19 - 20 5870 93
XM_017028673.1 XP_016884162.1 19 - 20 5664 93
GenBank mRNA AB028966.3 - 14 - 15 4484 93
AB665284.1 - 19 - 20 5639 93
BC016465.1 - 11 - 12 2235 93

View Full Product Details